Product Details

SNP ID
rs17009956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49365623 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAACAAGAGAAAGGGCATTACTGCT[C/T]ATCTTGGTCCAGGGGAGCAAGAGTC
Phenotype
MIM: 606701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRGX PubMed Links

Gene Details

Gene
DRGX
Gene Name
dorsal root ganglia homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276451.1 1471 Intron NP_001263380.1
XM_011540089.2 1471 UTR 3 XP_011538391.1

View Full Product Details