Product Details

SNP ID
rs16944363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60446097 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAAAATTCAACACACATCCATTCA[C/G]CCACAGACTACTCCAAGCATTAGCT
Phenotype
MIM: 605324
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APPBP2 PubMed Links

Gene Details

Gene
APPBP2
Gene Name
amyloid beta precursor protein binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282476.1 2763 UTR 3 NP_001269405.1
NM_006380.3 2763 UTR 3 NP_006371.2
XM_011524198.2 2763 UTR 3 XP_011522500.1
XM_011524199.2 2763 UTR 3 XP_011522501.1

View Full Product Details