Product Details

SNP ID
rs16891141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:33442972 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATACTAATAACCCCTGTGCTGTT[C/G]TGAGTGAGTCAGGGACAGATCATTC
Phenotype
MIM: 187790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TARS PubMed Links

Gene Details

Gene
TARS
Gene Name
threonyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258437.1 Intron NP_001245366.1
NM_001258438.1 Intron NP_001245367.1
NM_152295.4 Intron NP_689508.3

View Full Product Details