Product Details

SNP ID
rs17854483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50603290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCTTCCAGGATGACTTCCAGG[A/G]GTTTGAGATGATCGATGACAATGAA
Phenotype
MIM: 607755
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHKB-AS1 PubMed Links

Gene Details

Gene
CHKB-AS1
Gene Name
CHKB antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
MAPK8IP2
Gene Name
mitogen-activated protein kinase 8 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012324.4 262 Missense Mutation GAG,GGG E80G NP_036456.1
XM_011530679.2 262 Missense Mutation GAG,GGG E80G XP_011528981.1
XM_011530680.2 262 Missense Mutation GAG,GGG E80G XP_011528982.1
XM_011530681.2 262 Missense Mutation GAG,GGG E80G XP_011528983.1

View Full Product Details