Product Details
- SNP ID
-
rs17771103
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:87330772 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACCAACTTCCCGAGCTTGCTCTGC[A/G]TACTGTGCCCATCACACCTGCGCTG
- Phenotype
-
MIM: 609102
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO31
PubMed Links
Gene Details
- Gene
- FBXO31
- Gene Name
- F-box protein 31
View Full Product Details