Product Details

SNP ID
rs11541217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:131048679 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCTGCAGTATACTCAAGAGTCTG[C/T]TGCCCTTCAGAAAGCCAGAGGGAAC
Phenotype
MIM: 605216
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF4 PubMed Links

Gene Details

Gene
ARHGEF4
Gene Name
Rho guanine nucleotide exchange factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015320.3 4514 Intron NP_056135.2
NM_032995.2 4514 Intron NP_127462.1
Gene
FAM168B
Gene Name
family with sequence similarity 168 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009993.3 4514 UTR 3 NP_001009993.2
NM_001321743.1 4514 UTR 3 NP_001308672.1
NM_001321744.1 4514 UTR 3 NP_001308673.1
NM_001321745.1 4514 UTR 3 NP_001308674.1
NM_001321746.1 4514 UTR 3 NP_001308675.1
NM_001321747.1 4514 UTR 3 NP_001308676.1
NM_001321748.1 4514 UTR 3 NP_001308677.1
XM_011510594.2 4514 UTR 3 XP_011508896.1
XM_017003327.1 4514 UTR 3 XP_016858816.1
XM_017003328.1 4514 UTR 3 XP_016858817.1
Gene
POTEI
Gene Name
POTE ankyrin domain family member I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277406.1 4514 Intron NP_001264335.1
XM_011511670.2 4514 Intron XP_011509972.1
XM_017004732.1 4514 Intron XP_016860221.1
XM_017004733.1 4514 Intron XP_016860222.1
XM_017004734.1 4514 Intron XP_016860223.1
XM_017004735.1 4514 Intron XP_016860224.1

View Full Product Details