Product Details
- SNP ID
-
rs17426593
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:32640300 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTTAATAGTAGACCATGCCTGAT[C/T]GGTGTTTTACACATCTCCGGCTATG
- Phenotype
-
MIM: 146880
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HLA-DQA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28383404] are located under a probe and SNP(s) [rs28383403,rs28383405,rs74395243] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HLA-DQA1
- Gene Name
- major histocompatibility complex, class II, DQ alpha 1
View Full Product Details