Product Details

SNP ID
rs34298659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67893794 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGGCCGGCGGGGCAGCTGATCG[C/T]TTCCTTGTGCAGGCTGGGAAATAGC
Phenotype
MIM: 606030 MIM: 177015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EDC4 PubMed Links

Gene Details

Gene
EDC4
Gene Name
enhancer of mRNA decapping 4
There are no transcripts associated with this gene.

Gene
NRN1L
Gene Name
neuritin 1 like
There are no transcripts associated with this gene.

Gene
PSKH1
Gene Name
protein serine kinase H1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006742.2 Intron NP_006733.1

View Full Product Details