Product Details

SNP ID
rs17773563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69317126 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCTGGATGGAGGGTCTGCACCAC[A/G]TTCCCATCCGCAGAGGGAGAGGTCT
Phenotype
MIM: 600027 MIM: 609982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNTB2 PubMed Links

Gene Details

Gene
SNTB2
Gene Name
syntrophin beta 2
There are no transcripts associated with this gene.

Gene
VPS4A
Gene Name
vacuolar protein sorting 4 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013245.2 Intron NP_037377.1

View Full Product Details