Product Details
- SNP ID
-
rs17307260
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:16070725 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTGTTTCATGCCCTGTCTCATTT[C/T]GCATAGCTAAAAAAGAATGCTAATT
- Phenotype
-
MIM: 615964
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR99AHG
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199514533] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR99AHG
- Gene Name
- mir-99a-let-7c cluster host gene
There are no transcripts associated with this gene.
View Full Product Details