Product Details

SNP ID
rs16850073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73838282 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATATTCTCTTCCTATGGTTTTAGA[C/T]GTTTGATGTCTTCTTAGTATGGCAT
Phenotype
MIM: 138965
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCL6 PubMed Links

Gene Details

Gene
CXCL6
Gene Name
C-X-C motif chemokine ligand 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002993.3 Intron NP_002984.1

View Full Product Details