Product Details

SNP ID
rs16850898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74309960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGAGTCAGGCAAATAAAGGAAG[A/G]TTGTCATATCTCAGTGTTCTTCATG
Phenotype
MIM: 614047
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPGN PubMed Links

Gene Details

Gene
EPGN
Gene Name
epithelial mitogen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270989.1 Intron NP_001257918.1
NM_001270990.1 Intron NP_001257919.1
NM_001270991.1 Intron NP_001257920.1
NM_001270992.1 Intron NP_001257921.1
NM_001270993.1 Intron NP_001257922.1
XM_005265666.4 Intron XP_005265723.1
XM_011531817.2 Intron XP_011530119.1
Gene
LOC105377276
Gene Name
uncharacterized LOC105377276
There are no transcripts associated with this gene.

Gene
MTHFD2L
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
There are no transcripts associated with this gene.

View Full Product Details