Product Details

SNP ID
rs16982977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24310068 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTGAAAGAGGTTTCTCAGAGTT[C/T]ATAGGTCACTTTCATATATATTATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUPT20HL2 PubMed Links

Gene Details

Gene
SUPT20HL2
Gene Name
SPT20 homolog, SAGA complex component-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136233.1 Intron NP_001129705.1

View Full Product Details