Product Details
- SNP ID
-
hCV32898833
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:3058065 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCAAAGGTTTGTTCCTTTTCATTT[C/T]GTGTTCTAGTGTTTTCTTTTCCTGC
- Phenotype
-
MIM: 300003
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARSF
PubMed Links
Gene Details
- Gene
- ARSF
- Gene Name
- arylsulfatase F
There are no transcripts associated with this gene.
View Full Product Details