Product Details

SNP ID
rs17174697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:111905337 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGCATGTGAAACCCTGGGCTGCT[A/G]TGGCCAGGTCACCTCCTCAGGTCCT
Phenotype
MIM: 604705
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MERTK PubMed Links

Gene Details

Gene
MERTK
Gene Name
MER proto-oncogene, tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006343.2 Intron NP_006334.2
XM_005263565.4 Intron XP_005263622.1
XM_005263568.4 Intron XP_005263625.1
XM_011510490.2 Intron XP_011508792.1
XM_017003164.1 Intron XP_016858653.1
XM_017003165.1 Intron XP_016858654.1

View Full Product Details