Product Details

SNP ID
rs16835481
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:153659408 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGCTTTCCCTCAGAACAAGAGAT[A/G]AGCCGTTAGATCTTAGCTGCACCTT
Phenotype
MIM: 603181 MIM: 607007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ILF2 PubMed Links

Gene Details

Gene
ILF2
Gene Name
interleukin enhancer binding factor 2
There are no transcripts associated with this gene.

Gene
SNAPIN
Gene Name
SNAP associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012437.5 Intron NP_036569.1

View Full Product Details