Product Details

SNP ID
rs16931935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75027721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATGAACTTCACAATTATGGTTT[A/G]TTTCTAAGTGTCTTTAATATGCAAA
Phenotype
MIM: 605880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUPD1 PubMed Links

Gene Details

Gene
DUPD1
Gene Name
dual specificity phosphatase and pro isomerase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003892.1 Intron NP_001003892.1
XM_011539747.2 Intron XP_011538049.1
XM_017016176.1 Intron XP_016871665.1
Gene
KAT6B
Gene Name
lysine acetyltransferase 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256468.1 Intron NP_001243397.1
NM_001256469.1 Intron NP_001243398.1
NM_012330.3 Intron NP_036462.2
XM_005269664.2 Intron XP_005269721.1
XM_017016000.1 Intron XP_016871489.1
XM_017016001.1 Intron XP_016871490.1
XM_017016002.1 Intron XP_016871491.1
XM_017016003.1 Intron XP_016871492.1
XM_017016004.1 Intron XP_016871493.1
XM_017016005.1 Intron XP_016871494.1
XM_017016006.1 Intron XP_016871495.1
XM_017016007.1 Intron XP_016871496.1
XM_017016008.1 Intron XP_016871497.1
XM_017016009.1 Intron XP_016871498.1

View Full Product Details