Product Details

SNP ID
rs17136048
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6163064 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCTGACTTGCCTTTCTCAGAACTA[A/C]AACTTCCGATTTGAGGTATCAGTAA
Phenotype
MIM: 605081
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CYTH3 PubMed Links

Gene Details

Gene
CYTH3
Gene Name
cytohesin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004227.3 3184 UTR 3 NP_004218.1
Gene
USP42
Gene Name
ubiquitin specific peptidase 42
There are no transcripts associated with this gene.

View Full Product Details