Product Details
- SNP ID
-
rs17611220
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:26430004 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGGAACACATAGGACCAACGAAGA[A/G]ACCCTTTATCCTCCAGGACAGTGTG
- Phenotype
-
MIM: 613592
MIM: 613594
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BTN2A3P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75650916] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BTN2A3P
- Gene Name
- butyrophilin subfamily 2 member A3, pseudogene
There are no transcripts associated with this gene.
- Gene
- BTN3A2
- Gene Name
- butyrophilin subfamily 3 member A2
View Full Product Details