Product Details

SNP ID
rs17616564
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:108565773 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGCGGAAAGGATTTCTTTCTTTT[A/G]GCCAGAGTACATCCTTCTGTTTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM102B PubMed Links

Gene Details

Gene
FAM102B
Gene Name
family with sequence similarity 102 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010883.2 Intron NP_001010883.2

View Full Product Details