Product Details

SNP ID
rs17346310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97887251 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAATATTTCCAAAGTATGCTGGTA[C/T]CTTGTCTCCTCTAACTACACACACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRMO PubMed Links

Gene Details

Gene
TRMO
Gene Name
tRNA methyltransferase O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016481.4 Intron NP_057565.3
XM_005252047.3 Intron XP_005252104.1
XM_005252048.4 Intron XP_005252105.1
XM_005252049.4 Intron XP_005252106.1
XM_005252050.4 Intron XP_005252107.1
XM_011518776.2 Intron XP_011517078.1
XM_011518777.1 Intron XP_011517079.1

View Full Product Details