Product Details

SNP ID
rs17172871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44383229 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGTTAACAGCAGGTATGCACCT[C/G]GAGACCAAGGAGGGCCTTGGAGCCT
Phenotype
MIM: 610296
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUDCD3 PubMed Links

Gene Details

Gene
NUDCD3
Gene Name
NudC domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015332.3 4054 UTR 3 NP_056147.2
XM_011515247.2 4054 UTR 3 XP_011513549.1
XM_017011908.1 4054 Intron XP_016867397.1
XM_017011909.1 4054 UTR 3 XP_016867398.1
XM_017011910.1 4054 UTR 3 XP_016867399.1

View Full Product Details