Product Details
- SNP ID
-
rs17005366
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:69117205 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCTCACCCTGGCCACTGCTCCAG[A/G]AAGAATCAGAGCCTCTCTAAATCAT
- Phenotype
-
MIM: 616112
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LMOD3
PubMed Links
Gene Details
- Gene
- LMOD3
- Gene Name
- leiomodin 3
View Full Product Details