Product Details
- SNP ID
-
rs17853184
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:18283807 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGATTGACATACCATTGTCCCCTT[C/T]CATCAGGCTCATATCATTCAGATAC
- Phenotype
-
MIM: 607521
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HPS5
PubMed Links
Gene Details
- Gene
- HPS5
- Gene Name
- HPS5, biogenesis of lysosomal organelles complex 2 subunit 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007216.3 |
2980 |
Missense Mutation |
AAA,GAA |
K902E |
NP_009147.3 |
NM_181507.1 |
2980 |
Missense Mutation |
AAA,GAA |
K1016E |
NP_852608.1 |
NM_181508.1 |
2980 |
Missense Mutation |
AAA,GAA |
K902E |
NP_852609.1 |
XM_011519862.1 |
2980 |
Missense Mutation |
AAA,GAA |
K1016E |
XP_011518164.1 |
XM_011519863.1 |
2980 |
Missense Mutation |
AAA,GAA |
K1016E |
XP_011518165.1 |
XM_011519864.1 |
2980 |
Missense Mutation |
AAA,GAA |
K1016E |
XP_011518166.1 |
XM_011519865.1 |
2980 |
Missense Mutation |
AAA,GAA |
K979E |
XP_011518167.1 |
XM_011519866.1 |
2980 |
Missense Mutation |
AAA,GAA |
K902E |
XP_011518168.1 |
XM_011519867.1 |
2980 |
Missense Mutation |
AAA,GAA |
K902E |
XP_011518169.1 |
XM_011519868.2 |
2980 |
Missense Mutation |
AAA,GAA |
K902E |
XP_011518170.1 |
XM_017017149.1 |
2980 |
Missense Mutation |
AAA,GAA |
K1016E |
XP_016872638.1 |
XM_017017150.1 |
2980 |
Missense Mutation |
AAA,GAA |
K1016E |
XP_016872639.1 |
XM_017017151.1 |
2980 |
Missense Mutation |
AAA,GAA |
K979E |
XP_016872640.1 |
XM_017017152.1 |
2980 |
Missense Mutation |
AAA,GAA |
K979E |
XP_016872641.1 |
XM_017017153.1 |
2980 |
Missense Mutation |
AAA,GAA |
K979E |
XP_016872642.1 |
XM_017017154.1 |
2980 |
Missense Mutation |
AAA,GAA |
K902E |
XP_016872643.1 |
View Full Product Details