Product Details

SNP ID
rs17850140
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18269787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTCGCTGGCTGATCAGGCTGCC[A/G]ATGAATGGGGCAGGAGTGGCAAAGA
Phenotype
MIM: 607521 MIM: 104750
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HPS5 PubMed Links
Additional Information
For this assay, SNP(s) [rs12218,rs15790] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HPS5
Gene Name
HPS5, biogenesis of lysosomal organelles complex 2 subunit 2
There are no transcripts associated with this gene.

Gene
SAA1
Gene Name
serum amyloid A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000331.5 438 Missense Mutation AAT,GAT N101D NP_000322.2
NM_001178006.2 438 Missense Mutation AAT,GAT N101D NP_001171477.1
NM_199161.4 438 Missense Mutation AAT,GAT N101D NP_954630.1
Gene
ST13P5
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details