Product Details
- SNP ID
-
rs11558174
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:231208199 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAATGCAAAATAAAAGGAAAACCA[C/T]TGTGTAAAACAGTAGGCGGATCTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARMC9
PubMed Links
Gene Details
- Gene
- ARMC9
- Gene Name
- armadillo repeat containing 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001271466.2 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
NP_001258395.1 |
NM_001291656.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
NP_001278585.1 |
NM_025139.5 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
NP_079415.3 |
XM_011511905.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510207.1 |
XM_011511906.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510208.1 |
XM_011511907.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510209.1 |
XM_011511908.2 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510210.1 |
XM_011511909.2 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510211.1 |
XM_011511910.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510212.1 |
XM_011511911.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510213.1 |
XM_011511912.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510214.1 |
XM_011511913.2 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510215.1 |
XM_011511914.2 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510216.1 |
XM_011511915.2 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510217.1 |
XM_011511916.2 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_011510218.1 |
XM_011511919.2 |
163 |
Intron |
|
|
XP_011510221.1 |
XM_017005018.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860507.1 |
XM_017005019.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860508.1 |
XM_017005020.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860509.1 |
XM_017005021.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860510.1 |
XM_017005022.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860511.1 |
XM_017005023.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860512.1 |
XM_017005024.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860513.1 |
XM_017005025.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860514.1 |
XM_017005026.1 |
163 |
Silent Mutation |
CTG,TTG |
L42L |
XP_016860515.1 |
View Full Product Details