Product Details

SNP ID
rs16989998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45326973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCATGGTCAGTATGGGCTTGAGGG[C/T]GGACCTAGATTCTTAACTGGAAGTT
Phenotype
MIM: 616104 MIM: 600017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBPJL PubMed Links

Gene Details

Gene
RBPJL
Gene Name
recombination signal binding protein for immunoglobulin kappa J region like
There are no transcripts associated with this gene.

Gene
SDC4
Gene Name
syndecan 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002999.3 783 UTR 3 NP_002990.2
XM_011528977.2 783 UTR 3 XP_011527279.1

View Full Product Details