Product Details
- SNP ID
-
rs16989785
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45175491 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCAATTGCCAAGGTAATTCCCAGAG[C/G]CAGGATGGGGCTCAAGTCTCCTAGT
- Phenotype
-
MIM: 182257
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PI3
PubMed Links
Gene Details
- Gene
- PI3
- Gene Name
- peptidase inhibitor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002638.3 |
|
Intron |
|
|
NP_002629.1 |
View Full Product Details