Product Details

SNP ID
rs17102178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74716315 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCACAATTCAGAAAACACTTAGA[A/G]TGCTTCGAGTATCATTATGGACATC
Phenotype
MIM: 615380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AREL1 PubMed Links

Gene Details

Gene
AREL1
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.

Gene
FCF1
Gene Name
FCF1 rRNA-processing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318508.1 Intron NP_001305437.1
NM_015962.4 Intron NP_057046.1
XM_005267730.3 Intron XP_005267787.1
XM_011536815.2 Intron XP_011535117.1
XM_011536816.2 Intron XP_011535118.1

View Full Product Details