Product Details

SNP ID
rs17784271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:96283696 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTGAAAAGACACTTGTAAAAAAG[A/G]AAAAGCTTAAGGACATTTAATAATG
Phenotype
MIM: 616226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG2B PubMed Links

Gene Details

Gene
ATG2B
Gene Name
autophagy related 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006720187.2 8658 UTR 3 XP_006720250.1
XM_017021397.1 8658 Intron XP_016876886.1

View Full Product Details