Product Details

SNP ID
rs16979808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14740184 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCATGTTTGGTTGTATGATTGAGA[A/G]CATGGAGTTTACAATGTGTCATCAT
Phenotype
MIM: 606100 MIM: 611811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRE2 PubMed Links

Gene Details

Gene
ADGRE2
Gene Name
adhesion G protein-coupled receptor E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271052.1 Intron NP_001257981.1
NM_013447.3 Intron NP_038475.2
XM_011527948.2 Intron XP_011526250.1
XM_011527949.2 Intron XP_011526251.1
XM_011527951.2 Intron XP_011526253.1
XM_011527952.2 Intron XP_011526254.1
XM_011527953.2 Intron XP_011526255.1
XM_011527954.2 Intron XP_011526256.1
XM_011527955.2 Intron XP_011526257.1
XM_017026726.1 Intron XP_016882215.1
XM_017026727.1 Intron XP_016882216.1
Gene
ZNF333
Gene Name
zinc finger protein 333
There are no transcripts associated with this gene.

View Full Product Details