Product Details

SNP ID
rs16933138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66989372 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTTTTCCTATCCAGTGTCACTC[C/T]TTCCTCAGTACTTTGTCTGTTATGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R42 PubMed Links

Gene Details

Gene
PPP1R42
Gene Name
protein phosphatase 1 regulatory subunit 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013626.2 Intron NP_001013648.1

View Full Product Details