Product Details

SNP ID
rs16938727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73789533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAATTTATGTTGAGAAAGCAAGTC[A/T]CCCACAAATTATTTTTAAAAAATGA
Phenotype
MIM: 614277
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2W PubMed Links

Gene Details

Gene
UBE2W
Gene Name
ubiquitin conjugating enzyme E2 W (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001481.2 5146 UTR 3 NP_001001481.2
NM_001271015.1 5146 Intron NP_001257944.1
NM_018299.4 5146 UTR 3 NP_060769.4

View Full Product Details