Product Details

SNP ID
rs16846420
Assay Type
Functionally Tested
NCBI dbSNP Submissions
35
Location
Chr.1:173803368 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCTGTGTTAGACTCTGCTCCATT[G/T]GCAAGGAATAAGGGCAGACAGGTGA
Phenotype
MIM: 611503
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
CENPL PubMed Links

Gene Details

Gene
CENPL
Gene Name
centromere protein L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127181.2 1161 Silent Mutation GCA,GCC A232A NP_001120653.1
NM_001171182.1 1161 Silent Mutation GCA,GCC A186A NP_001164653.1
NM_033319.3 1161 Silent Mutation GCA,GCC A186A NP_201576.1

View Full Product Details