Product Details

SNP ID
rs17163788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:10938003 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTCAATATAATGGAATTTTACTC[A/G]TATATCCAGTTTCAAGAAAACCCAC
Phenotype
MIM: 603833
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFA4 PubMed Links

Gene Details

Gene
NDUFA4
Gene Name
NDUFA4, mitochondrial complex associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002489.3 Intron NP_002480.1

View Full Product Details