Product Details

SNP ID
rs16947838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64513842 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCATTTAGAATAGGATACCACA[A/G]TCTGAAAGTGTTCTCTCCTGCCACT
Phenotype
MIM: 180630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP95 PubMed Links

Gene Details

Gene
CEP95
Gene Name
centrosomal protein 95
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316990.1 Intron NP_001303919.1
NM_138363.2 Intron NP_612372.1
XM_005257779.3 Intron XP_005257836.1
XM_005257780.2 Intron XP_005257837.1
XM_006722166.3 Intron XP_006722229.1
XM_011525458.2 Intron XP_011523760.1
XM_017025287.1 Intron XP_016880776.1
XM_017025288.1 Intron XP_016880777.1
Gene
DDX5
Gene Name
DEAD-box helicase 5
There are no transcripts associated with this gene.

View Full Product Details