Product Details

SNP ID
rs13405145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119445102 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTCTCTTGGCAAGACATTTTCC[C/T]AGAGAAGTAGTCCACAGTCCCAACC
Phenotype
MIM: 182098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCTR PubMed Links

Gene Details

Gene
SCTR
Gene Name
secretin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002980.2 Intron NP_002971.2
XM_005263730.4 Intron XP_005263787.1
XM_011511621.2 Intron XP_011509923.1
XM_017004670.1 Intron XP_016860159.1
XM_017004671.1 Intron XP_016860160.1
XM_017004672.1 Intron XP_016860161.1
XM_017004673.1 Intron XP_016860162.1
Gene
TMEM37
Gene Name
transmembrane protein 37
There are no transcripts associated with this gene.

View Full Product Details