Product Details

SNP ID
rs17024799
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:95868587 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAGTTCTTACCTCTTGAGGTTAG[C/T]TGTAAGGCACACAGTAAGCATTCGT
Phenotype
MIM: 603541
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC38 PubMed Links

Gene Details

Gene
CCDC38
Gene Name
coiled-coil domain containing 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182496.2 Intron NP_872302.2
XM_006719229.2 Intron XP_006719292.1
XM_011537883.2 Intron XP_011536185.1
XM_011537884.2 Intron XP_011536186.1
XM_011537886.2 Intron XP_011536188.1
XM_011537887.2 Intron XP_011536189.1
XM_011537888.2 Intron XP_011536190.1
XM_011537889.1 Intron XP_011536191.1
Gene
LOC105369921
Gene Name
uncharacterized LOC105369921
There are no transcripts associated with this gene.

Gene
SNRPF
Gene Name
small nuclear ribonucleoprotein polypeptide F
There are no transcripts associated with this gene.

View Full Product Details