Product Details
- SNP ID
-
rs16857588
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:11535020 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAGCGGCTCTAGTTCTTGTTGGTT[C/G]ATTCAATTTGTATCAGAAGAGCTTT
- Phenotype
-
MIM: 611736
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GREB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114688802] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GREB1
- Gene Name
- growth regulation by estrogen in breast cancer 1
- Gene
- MIR4429
- Gene Name
- microRNA 4429
There are no transcripts associated with this gene.
View Full Product Details