Product Details

SNP ID
rs17041739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:221492 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGGTCCCTGCCCTCACAATCAG[G/T]TGTTGGTCTAATTTTCTCTGAATTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SH3YL1 PubMed Links

Gene Details

Gene
SH3YL1
Gene Name
SH3 and SYLF domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159597.2 Intron NP_001153069.1
NM_001282682.1 Intron NP_001269611.1
NM_001282687.1 Intron NP_001269616.1
NM_015677.3 Intron NP_056492.2

View Full Product Details