Product Details

SNP ID
rs17064587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:177309917 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTGCACAGCGGTATTCTCACCAG[A/G]CCCTGCAATCGGTGGGCCACAGTGC
Phenotype
MIM: 608934
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEIL3 PubMed Links

Gene Details

Gene
NEIL3
Gene Name
nei like DNA glycosylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018248.2 47 UTR 5 NP_060718.2
XM_017008360.1 47 UTR 5 XP_016863849.1

View Full Product Details