Product Details

SNP ID
rs17001550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76161120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTTGATTCTACCCAGTATTTTA[C/T]TGCAGTTGGAAGACTGAGTTTTCCT
Phenotype
MIM: 602257
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCARB2 PubMed Links

Gene Details

Gene
SCARB2
Gene Name
scavenger receptor class B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204255.1 1957 UTR 3 NP_001191184.1
NM_005506.3 1957 UTR 3 NP_005497.1

View Full Product Details