Product Details

SNP ID
rs17097400
Assay Type
Functionally Tested
NCBI dbSNP Submissions
33
Location
Chr.1:68474883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAAAAGATAGTTAATAAGGCTC[A/C]TCAATTGTCCGTAATTTTGTTTTGG
Phenotype
MIM: 612002
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DEPDC1 PubMed Links

Gene Details

Gene
DEPDC1
Gene Name
DEP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114120.2 4707 UTR 3 NP_001107592.1
NM_017779.5 4707 UTR 3 NP_060249.2

View Full Product Details