Product Details

SNP ID
rs17080221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118463222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAAACAAAAACGTATCTATAGT[A/G]GATTAAAACAAACTGTAACGTCTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP85L PubMed Links

Gene Details

Gene
CEP85L
Gene Name
centrosomal protein 85 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042475.2 5780 UTR 3 NP_001035940.1
NM_001178035.1 5780 UTR 3 NP_001171506.1
NM_206921.2 5780 Intron NP_996804.2
XM_005266970.1 5780 UTR 3 XP_005267027.1
XM_005266971.1 5780 UTR 3 XP_005267028.1
XM_005266972.4 5780 UTR 3 XP_005267029.1
XM_006715475.3 5780 UTR 3 XP_006715538.1
XM_011535809.2 5780 UTR 3 XP_011534111.1
XM_011535810.2 5780 UTR 3 XP_011534112.1
XM_011535811.1 5780 UTR 3 XP_011534113.1
XM_017010846.1 5780 UTR 3 XP_016866335.1
XM_017010847.1 5780 UTR 3 XP_016866336.1

View Full Product Details