Product Details
- SNP ID
-
rs17511525
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:111307568 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCGCGAGGAGCTGGTAGCTTTATG[G/T]ATCTATAAAAACATCCATTCAGAAG
- Phenotype
-
MIM: 602776
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
REV3L
PubMed Links
Gene Details
- Gene
- REV3L
- Gene Name
- REV3 like, DNA directed polymerase zeta catalytic subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286431.1 |
9675 |
Silent Mutation |
ATA,ATC |
I2937I |
NP_001273360.1 |
NM_001286432.1 |
9675 |
Silent Mutation |
ATA,ATC |
I2937I |
NP_001273361.1 |
NM_002912.4 |
9675 |
Silent Mutation |
ATA,ATC |
I3015I |
NP_002903.3 |
XM_011536028.1 |
9675 |
Silent Mutation |
ATA,ATC |
I3042I |
XP_011534330.1 |
XM_011536029.2 |
9675 |
Silent Mutation |
ATA,ATC |
I3041I |
XP_011534331.1 |
XM_011536030.2 |
9675 |
Silent Mutation |
ATA,ATC |
I3016I |
XP_011534332.1 |
XM_011536031.2 |
9675 |
Silent Mutation |
ATA,ATC |
I2964I |
XP_011534333.1 |
XM_011536032.2 |
9675 |
Silent Mutation |
ATA,ATC |
I2964I |
XP_011534334.1 |
XM_011536036.2 |
9675 |
Intron |
|
|
XP_011534338.1 |
XM_017011152.1 |
9675 |
Silent Mutation |
ATA,ATC |
I2963I |
XP_016866641.1 |
XM_017011153.1 |
9675 |
Silent Mutation |
ATA,ATC |
I2963I |
XP_016866642.1 |
XM_017011154.1 |
9675 |
Silent Mutation |
ATA,ATC |
I2963I |
XP_016866643.1 |
XM_017011155.1 |
9675 |
Intron |
|
|
XP_016866644.1 |
View Full Product Details