Product Details

SNP ID
rs16861729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187217538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAATTAATATTCACAATCAGTGA[C/T]TCAAAAAGCAGCAATTTTAAAGTTG
Phenotype
MIM: 600521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929106 PubMed Links

Gene Details

Gene
LOC101929106
Gene Name
uncharacterized LOC101929106
There are no transcripts associated with this gene.

Gene
MASP1
Gene Name
mannan binding lectin serine peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031849.2 5023 Intron NP_001027019.1
NM_001879.5 5023 UTR 3 NP_001870.3
NM_139125.3 5023 Intron NP_624302.1
XM_006713701.2 5023 Intron XP_006713764.1
XM_011512989.2 5023 Intron XP_011511291.1
XM_011512990.2 5023 Intron XP_011511292.1
XM_011512991.2 5023 Intron XP_011511293.1
XM_017006869.1 5023 Intron XP_016862358.1
XM_017006870.1 5023 Intron XP_016862359.1
XM_017006871.1 5023 Intron XP_016862360.1
XM_017006872.1 5023 Intron XP_016862361.1

View Full Product Details