Product Details

SNP ID
rs16837708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126711405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTTGCCTCCTTGACTGTTTCAG[C/T]TTGAACATGGAAGTTCCTTATGATA
Phenotype
MIM: 615634
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD6 PubMed Links

Gene Details

Gene
CHCHD6
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320610.1 Intron NP_001307539.1
NM_032343.2 Intron NP_115719.1
XM_011513234.1 Intron XP_011511536.1
XM_011513235.2 Intron XP_011511537.1
XM_017007320.1 Intron XP_016862809.1
XM_017007321.1 Intron XP_016862810.1
XM_017007322.1 Intron XP_016862811.1

View Full Product Details