Product Details

SNP ID
rs17108480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:70639503 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGTTCCCAGCACACACTAGGCA[C/T]GTGATTAATTTTTACCTGTGTAACA
Phenotype
MIM: 176882 MIM: 602853
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPRB PubMed Links

Gene Details

Gene
PTPRB
Gene Name
protein tyrosine phosphatase, receptor type B
There are no transcripts associated with this gene.

Gene
PTPRR
Gene Name
protein tyrosine phosphatase, receptor type R
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207015.1 Intron NP_001193944.1
NM_001207016.1 Intron NP_001193945.1
NM_002849.3 Intron NP_002840.2
NM_130846.2 Intron NP_570897.2
XM_011538615.2 Intron XP_011536917.1

View Full Product Details