Product Details

SNP ID
rs17041076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:92737887 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTATTACTCTACTGTCTCTTTAT[A/G]CTTTAACATTGGATGAGATCGTCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHG7 PubMed Links

Gene Details

Gene
PLEKHG7
Gene Name
pleckstrin homology and RhoGEF domain containing G7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004330.2 Intron NP_001004330.1

View Full Product Details