Product Details

SNP ID
rs17020720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:92773036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTAAGACACAATAATGTGTGGCA[C/T]GTTATAATTACAGAATACCATTCAT
Phenotype
MIM: 605070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEA1 PubMed Links

Gene Details

Gene
EEA1
Gene Name
early endosome antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003566.3 7476 UTR 3 NP_003557.2
XM_011538814.2 7476 UTR 3 XP_011537116.1
XM_011538815.2 7476 UTR 3 XP_011537117.1
XM_017020018.1 7476 UTR 3 XP_016875507.1
Gene
PLEKHG7
Gene Name
pleckstrin homology and RhoGEF domain containing G7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004330.2 7476 Intron NP_001004330.1

View Full Product Details